siRNA Details
| Gene Symbol: 1700065O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045386.1 | 1 | 22 | |||
| GenBank mRNA | AK006898.1 | - | 1 | 25 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700065O20Rik| Gene Symbol: 1700065O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045386.1 | 1 | 22 | |||
| GenBank mRNA | AK006898.1 | - | 1 | 25 | ||