siRNA Details
| Gene Symbol: 1700065O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045386.1 | 4, 5 | 511 | |||
| GenBank mRNA | AK006898.1 | - | 4, 5 | 514 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706891.1 | - | 4, 5 | 513 | Chr. 18 - 49803326 - 49817958 [-] on Build GRCm38 | |

1700065O20Rik
Genomic Map