siRNA Details
| Gene Symbol: Kis2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY940617.1 | - | LIT3390 Kis2 |
1, 2 | 798 | |
| AY940618.1 | - | LIT3391 Kis2 |
2, 3 | 484 | ||

Kis2
Genomic Map