siRNA Details
| Gene Symbol: 2310067P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010100.1 | - | 1 | 237 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709636.1 | - | 1 | 237 | Chr. 13 - 83065457 - 83194259 [+] on Build GRCm38 | |

2310067P03Rik
Genomic Map