siRNA Details
Gene Symbol: 4930523O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040451.1 | 8 | 742 | |||
GenBank mRNA | AK014957.1 | - | 8 | 1076 | ||
AK015877.1 | - | 8 | 744 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930523O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040451.1 | 8 | 742 | |||
GenBank mRNA | AK014957.1 | - | 8 | 1076 | ||
AK015877.1 | - | 8 | 744 |