siRNA Details
| Gene Symbol: 4930523O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040451.1 | 2 | 141 | |||
| GenBank mRNA | AK014957.1 | - | 2 | 254 | ||
| AK015877.1 | - | 2 | 143 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930523O13Rik| Gene Symbol: 4930523O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040451.1 | 2 | 141 | |||
| GenBank mRNA | AK014957.1 | - | 2 | 254 | ||
| AK015877.1 | - | 2 | 143 | |||