siRNA Details
Gene Symbol: 4921531C22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033782.1 | 2 | 597 | |||
GenBank mRNA | AK014955.1 | - | 2 | 547 | ||
AK030577.1 | - | 2 | 568 | |||
AK076601.1 | - | 2 | 611 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4921531C22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033782.1 | 2 | 597 | |||
GenBank mRNA | AK014955.1 | - | 2 | 547 | ||
AK030577.1 | - | 2 | 568 | |||
AK076601.1 | - | 2 | 611 |