siRNA Details
Gene Symbol: 4930474H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045712.1 | 8 | 1339 | |||
GenBank mRNA | AK015575.1 | - | 8 | 1346 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930474H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045712.1 | 8 | 1339 | |||
GenBank mRNA | AK015575.1 | - | 8 | 1346 |