siRNA Details
Gene Symbol: Gm13856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015572.1 | - | 267 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715122.1 | - | 2 | 267 | Chr. 6 - 34151929 - 34163945 [+] on Build GRCm38 |
