siRNA Details
| Gene Symbol: Gm13856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015572.1 | - | 138 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715122.1 | - | 2 | 138 | Chr. 6 - 34151929 - 34163945 [+] on Build GRCm38 | |

Gm13856
Genomic Map