siRNA Details
Gene Symbol: 4930474B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015571.1 | - | 1 | 415 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715121.1 | - | 1 | 415 | Chr. 1 - 90870591 - 90871419 [-] on Build GRCm38 |
