siRNA Details
| Gene Symbol: Gm35387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC066010.1 | - | 2 | 2403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm35387| Gene Symbol: Gm35387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC066010.1 | - | 2 | 2403 | ||