siRNA Details
| Gene Symbol: 4930477E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015579.1 | - | 2 | 384 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715128.1 | - | 2 | 384 | Chr. 3 - 96401167 - 96402014 [-] on Build GRCm38 | |

4930477E14Rik
Genomic Map