siRNA Details
| Gene Symbol: 3010001F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031183.1 | - | 1 | 1382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3010001F23Rik| Gene Symbol: 3010001F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031183.1 | - | 1 | 1382 | ||