siRNA Details
| Gene Symbol: G630018N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137190.1 | - | 3 | 1589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G630018N14Rik| Gene Symbol: G630018N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137190.1 | - | 3 | 1589 | ||