siRNA Details
| Gene Symbol: 1810013A23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045427.1 | 3 | 404 | |||
| GenBank mRNA | AK007468.1 | - | 3 | 405 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707535.1 | - | 3 | 405 | Chr. 17 - 28271708 - 28273163 [+] on Build GRCm38 | |

1810013A23Rik
Genomic Map