siRNA Details
| Gene Symbol: 4933424C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016884.1 | - | 1 | 50 | ||
| AK048826.1 | - | 1 | 171 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933424C08Rik| Gene Symbol: 4933424C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016884.1 | - | 1 | 50 | ||
| AK048826.1 | - | 1 | 171 | |||