siRNA Details
| Gene Symbol: 4933424C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016884.1 | - | 1 | 772 | ||
| AK048826.1 | - | 1 | 893 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB391148.2 | - | 1 | 283 | Chr. 4 - 126968743 - 126969379 [+] on Build GRCm38 | |

4933424C08Rik
Genomic Map