siRNA Details
| Gene Symbol: 1700109G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033788.1 | 3 | 682 | |||
| GenBank mRNA | AK007149.1 | - | 3 | 682 | ||
| AK018935.1 | - | 3 | 645 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU935660.1 | - | 3 | 638 | Chr. 14 - 61292980 - 61305289 [-] on Build GRCm38 | |

1700109G14Rik
Genomic Map