siRNA Details
| Gene Symbol: Carlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017121.1 | - | 1 | 1393 | ||
| AK018938.1 | - | 1 | 134 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Carlr| Gene Symbol: Carlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017121.1 | - | 1 | 1393 | ||
| AK018938.1 | - | 1 | 134 | |||