siRNA Details
| Gene Symbol: 2700070H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046019.1 | 3 | 485 | |||
| GenBank mRNA | AK012509.1 | - | 3 | 485 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711634.1 | - | 3 | 488 | Chr. 14 - 63055073 - 63056940 [+] on Build GRCm38 | |

2700070H01Rik
Genomic Map