siRNA Details
| Gene Symbol: 2700070H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046019.1 | 3 | 380 | |||
| GenBank mRNA | AK012509.1 | - | 3 | 380 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2700070H01Rik| Gene Symbol: 2700070H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046019.1 | 3 | 380 | |||
| GenBank mRNA | AK012509.1 | - | 3 | 380 | ||