siRNA Details
| Gene Symbol: 4930550C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016084.1 | - | 448 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715686.1 | - | 4 | 452 | Chr. 13 - 56222836 - 56243940 [-] on Build GRCm38 | |

4930550C17Rik
Genomic Map