siRNA Details
| Gene Symbol: Top2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052306.1 | - | 1750 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB477439.2 | - | 1 | 420 | Chr. 14 - 16431986 - 16432638 [+] on Build GRCm38 | |

Top2b
Genomic Map