siRNA Details
| Gene Symbol: Wwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI847908.1 | - | 1 | 382 | ||
| AK038924.1 | - | 927 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB178800.2 | - | 1 | 379 | Chr. 4 - 19607796 - 19608409 [+] on Build GRCm38 | |

Wwp1
Genomic Map