siRNA Details
Gene Symbol: 1700028M03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036591.1 | 3 | 314 | |||
GenBank mRNA | AK006465.1 | - | 3 | 327 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700028M03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036591.1 | 3 | 314 | |||
GenBank mRNA | AK006465.1 | - | 3 | 327 |