siRNA Details
| Gene Symbol: Gm29686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084744.1 | - | 2 | 510 | ||
| AK157531.1 | - | 512 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm29686| Gene Symbol: Gm29686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084744.1 | - | 2 | 510 | ||
| AK157531.1 | - | 512 | ||||