siRNA Details
| Gene Symbol: 2310021N16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009452.1 | - | 1 | 467 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709178.1 | - | 1 | 466 | Chr. 2 - 148280475 - 148281157 [-] on Build GRCm38 | |

2310021N16Rik
Genomic Map