siRNA Details
| Gene Symbol: Gm12216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039036.1 | - | 3 | 343 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB632853.1 | - | 3 | 343 | Chr. 11 - 53812948 - 53850077 [-] on Build GRCm38 | |

Gm12216
Genomic Map