siRNA Details
| Gene Symbol: Sc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050439.1 | - | 1 | 2775 | ||
| AK080673.1 | - | 1 | 1425 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB667866.1 | - | 1 | 629 | Chr. 9 - 42251598 - 42252292 [-] on Build GRCm38 | |

Sc5d
Genomic Map