siRNA Details
| Gene Symbol: 4933440J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045344.1 | 3 | 372 | |||
| GenBank mRNA | AK017133.1 | - | 3 | 373 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716812.1 | - | 3 | 373 | Chr. 10 - 111586917 - 111594273 [-] on Build GRCm38 | |

4933440J02Rik
Genomic Map