siRNA Details
| Gene Symbol: Ddi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018643.1 | - | 1614 | |||
| AK078461.1 | - | 1986 | ||||
| AK165215.1 | - | 1375 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddi2| Gene Symbol: Ddi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018643.1 | - | 1614 | |||
| AK078461.1 | - | 1986 | ||||
| AK165215.1 | - | 1375 | ||||