siRNA Details
| Gene Symbol: A330087I24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039689.1 | - | 8 | 2610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330087I24| Gene Symbol: A330087I24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039689.1 | - | 8 | 2610 | ||