siRNA Details
| Gene Symbol: Tox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK169546.1 | - | 4 | 3403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tox| Gene Symbol: Tox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK169546.1 | - | 4 | 3403 | ||