siRNA Details
| Gene Symbol: 4933402J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040682.1 | 1 | 343 | |||
| NR_040683.1 | 1 | 343 | ||||
| GenBank mRNA | AK016620.1 | - | 1 | 346 | ||
| BC099582.1 | - | 1 | 339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933402J10Rik| Gene Symbol: 4933402J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040682.1 | 1 | 343 | |||
| NR_040683.1 | 1 | 343 | ||||
| GenBank mRNA | AK016620.1 | - | 1 | 346 | ||
| BC099582.1 | - | 1 | 339 | |||