siRNA Details
Gene Symbol: 4933402J10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040682.1 | 1 | 613 | |||
NR_040683.1 | 1 | 613 | ||||
GenBank mRNA | AK016620.1 | - | 1 | 616 | ||
BC099582.1 | - | 1 | 609 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933402J10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040682.1 | 1 | 613 | |||
NR_040683.1 | 1 | 613 | ||||
GenBank mRNA | AK016620.1 | - | 1 | 616 | ||
BC099582.1 | - | 1 | 609 |