siRNA Details
| Gene Symbol: 4632404H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035453.1 | - | 1 | 240 | ||
| BC063049.1 | - | 1 | 2816 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB627495.1 | - | 1 | 240 | Chr. 3 - 89769598 - 89770215 [-] on Build GRCm38 | |

4632404H12Rik
Genomic Map