siRNA Details
| Gene Symbol: 4632404H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014549.2 | - | 1708 | |||
| AK035453.1 | - | 1 | 1623 | |||
| BC063049.1 | - | 1 | 4199 | |||
| BC106127.1 | - | 2 | 1715 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4632404H12Rik| Gene Symbol: 4632404H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014549.2 | - | 1708 | |||
| AK035453.1 | - | 1 | 1623 | |||
| BC063049.1 | - | 1 | 4199 | |||
| BC106127.1 | - | 2 | 1715 | |||