siRNA Details
Gene Symbol: 4632404H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014549.2 | - | 1071 | |||
AK035453.1 | - | 1 | 984 | |||
BC063049.1 | - | 1 | 3561 | |||
BC106127.1 | - | 2 | 1077 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4632404H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014549.2 | - | 1071 | |||
AK035453.1 | - | 1 | 984 | |||
BC063049.1 | - | 1 | 3561 | |||
BC106127.1 | - | 2 | 1077 |