siRNA Details
| Gene Symbol: Uba6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017687.1 | - | 976 | |||
| AK029740.1 | - | 2260 | ||||
| AK139327.1 | - | 1418 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uba6| Gene Symbol: Uba6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017687.1 | - | 976 | |||
| AK029740.1 | - | 2260 | ||||
| AK139327.1 | - | 1418 | ||||