siRNA Details
| Gene Symbol: Gpkow | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017864.1 | - | 1 | 474 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA527122.1 | - | 1 | 150 | Chr. X - 7697499 - 7698098 [+] on Build GRCm38 | |

Gpkow
Genomic Map