siRNA Details
| Gene Symbol: Klhdc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK157991.1 | - | 1 | 1269 | ||
| AK161517.1 | - | 1 | 418 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ047877.1 | - | 1 | 418 | Chr. 9 - 108459240 - 108460078 [-] on Build GRCm38 | |

Klhdc8b
Genomic Map