siRNA Details
| Gene Symbol: Gm30223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083219.1 | - | 2 | 122 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB653349.1 | - | 2 | 122 | Chr. 3 - 16817092 - 16824180 [+] on Build GRCm38 | |

Gm30223
Genomic Map