siRNA Details
| Gene Symbol: Iqcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083217.1 | - | 1 | 687 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ132188.1 | - | 2 | 278 | Chr. 4 - 129618243 - 129619080 [-] on Build GRCm38 | |

Iqcc
Genomic Map