siRNA Details
| Gene Symbol: 2900084I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013819.1 | - | 1 | 88 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712784.1 | - | 1 | 56 | Chr. 6 - 126138690 - 126139191 [-] on Build GRCm38 | |

2900084I15Rik
Genomic Map