siRNA Details
| Gene Symbol: 4833445I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076537.1 | - | 1 | 1378 | ||
| AV253134.2 | - | 1 | 388 | |||
| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK217499.1 | - | 1 | 199 | ||

4833445I07Rik
Genomic Map