siRNA Details
| Gene Symbol: 6030443J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044020.1 | - | 1334 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB272965.2 | - | 2 | 629 | Chr. 5 - 22694838 - 22701499 [+] on Build GRCm38 | |

6030443J06Rik
Genomic Map