siRNA Details
| Gene Symbol: 6030443J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044020.1 | - | 465 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB641926.1 | - | 6 | 463 | Chr. 5 - 22550437 - 22694768 [+] on Build GRCm38 | |

6030443J06Rik
Genomic Map