siRNA Details
| Gene Symbol: 6720454L07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020125.1 | - | 1 | 924 | ||
| AK137081.1 | - | 1 | 1736 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6720454L07Rik| Gene Symbol: 6720454L07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020125.1 | - | 1 | 924 | ||
| AK137081.1 | - | 1 | 1736 | |||