siRNA Details
| Gene Symbol: Gnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020124.1 | - | 1 | 746 | ||
| AK051397.1 | - | 1 | 187 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF744724.1 | - | 1 | 792 | Chr. 3 - 32582848 - 32583659 [-] on Build GRCm38 | |

Gnb4
Genomic Map